Anti-VEGFA Rabbit Recombinant Antibody [clone: VEGFA/7758R]
Catalog # 77181-240
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Folliculostellate cell-derived growth factor
- Antigen Symbol:VEGFA
- Clonality:Recombinant
- Clone:VEGFA/7758R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P15692
- Antigen Synonyms:vascular endothelial growth factor|MVCD1|vascular endothelial growth factor A165|MGC70609|Vegf|Vascular permeability factor|Glioma-derived endothelial cell mitogen|Vascular endothelial growth factor A|vascular endothelial growth factor A121
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:19 - 22 kDa (reducing)/38 kDa - 44 kDa (non - reducing)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human VEGFA protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77181-240
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [VEGFA/7758R] antibody to VEGFA for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: VEGFA
Clonality: Recombinant
Clone: VEGFA/7758R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Human