Anti-Transcription factor AP-2 alpha Mouse Monoclonal Antibody [clone: PCRP-TFAP2A-2C2]
Catalog # 77181-168
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Transcription factor AP-2 alpha
- Antigen Symbol:Transcription factor AP-2 alpha
- Clonality:Monoclonal
- Clone:PCRP-TFAP2A-2C2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05549
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:48 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human TFAP2A protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77181-168
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-TFAP2A-2C2] antibody to Transcription factor AP-2 alpha for Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Transcription factor AP-2 alpha
Clonality: Monoclonal
Clone: PCRP-TFAP2A-2C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human