Anti-TIMP2 Rabbit Recombinant Antibody [clone: TIMP2/8193R]
77181-148
- Antibody Type:Primary
- Antigen Name:CSC 21K
- Antigen Symbol:TIMP2
- Clonality:Recombinant
- Clone:TIMP2/8193R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P16035
- Antigen Synonyms:DDC8|MGC105282|OTTMUSP00000004171|CSC21K|CSC-21K|Metalloproteinase inhibitor 2 precursor|OTTMUSP00000004170|D11Bwg1104e|Metalloproteinase inhibitor 2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:21 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human TIMP2 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77181-148
- Packaging:Plastic vial
Recombinant rabbit monoclonal [TIMP2/8193R] antibody to TIMP2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: TIMP2
Clonality: Recombinant
Clone: TIMP2/8193R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human