Anti-TIMP1 Mouse Monoclonal Antibody [clone: TIMP1/4358]
77181-146
- Antibody Type:Primary
- Antigen Name:Clgi
- Antigen Symbol:TIMP1
- Clonality:Monoclonal
- Clone:TIMP1/4358
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01033
- Antigen Synonyms:Collagenase inhibitor, Human|Erythroid Potentiating Activity|Fibroblast collagenase inhibitor|EPO|FLJ90373|HCI|Collagenase inhibitor|EPA|Erythroid-potentiating activity
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:23 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human TIMP1 protein fragment (around Amino Acids 1-200). The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:77181-146
- Packaging:Plastic vial
Mouse monoclonal [TIMP1/4358] antibody to TIMP1 for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: ELISA: 2 to 4 µg/ ml for coating, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: TIMP1
Clonality: Monoclonal
Clone: TIMP1/4358
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human