Anti-Somatostatin Receptor 2 Mouse Monoclonal Antibody [clone: SSTR2/7532]
77181-018
- Antibody Type:Primary
- Antigen Name:Somatostatin Receptor 2
- Antigen Symbol:Somatostatin Receptor 2
- Clonality:Monoclonal
- Clone:SSTR2/7532
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b lambda
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P30874
- Antigen Synonyms:SRIF1|Somatostatin receptor type 2|SSTR2|SST2|SS2-R|SSR2_HUMAN|SS2R|SS-2-R|SRIF-1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:87 kDa/148 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 300-500) of human SSTR2. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77181-018
- Packaging:Plastic vial
Mouse monoclonal [SSTR2/7532] antibody to Somatostatin Receptor 2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Somatostatin Receptor 2
Clonality: Monoclonal
Clone: SSTR2/7532
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b lambda
Reactivity: Human