Anti-smooth muscle Myosin heavy chain 11 Mouse Monoclonal Antibody [clone: SM-M10]
77180-996
- Antibody Type:Primary
- Antigen Name:smooth muscle Myosin heavy chain 11
- Antigen Symbol:smooth muscle Myosin heavy chain 11
- Clonality:Monoclonal
- Clone:SM-M10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35749
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:205 kDa (MHC-1)/200 kDa (MHC-2)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Crude human uterus extract.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-996
- Packaging:Plastic vial
Mouse monoclonal [SM-M10] antibody to smooth muscle Myosin heavy chain 11 for Flow Cytometry, IF and IHC-P with samples derived from Human, Bovine and Mouse.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 0.5 to 1 µg/million cells, IF: 0.5 to 1 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: smooth muscle Myosin heavy chain 11
Clonality: Monoclonal
Clone: SM-M10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Bovine;Mouse