Anti-S100P Rabbit Recombinant Antibody [clone: S100P/7978R]
Catalog # 77180-876
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:MIG9
- Antigen Symbol:S100P
- Clonality:Recombinant
- Clone:S100P/7978R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P25815
- Antigen Synonyms:S100P_HUMAN|Migration inducing gene 9|Protein S100-P|S100 calcium-binding protein P|Protein S100-E|Protein S100P|S100 calcium binding protein P|S100E|S100 P
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:10 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-95) of human MIG9 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-876
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [S100P/7978R] antibody to S100P for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: S100P
Clonality: Recombinant
Clone: S100P/7978R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human