Anti-S100A14 Mouse Monoclonal Antibody [clone: S100A14/7401]
Catalog # 77180-832
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BCMP84
- Antigen Symbol:S100A14
- Clonality:Monoclonal
- Clone:S100A14/7401
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9HCY8
- Antigen Synonyms:Protein S100-A14|S114|S100A15|S100 calcium binding protein A14|S10AE_HUMAN|S100a14|S100 calcium-binding protein A14
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:11 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human S100A14 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-832
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [S100A14/7401] antibody to S100A14 for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: S100A14
Clonality: Monoclonal
Clone: S100A14/7401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human