Anti-RHOBTB2 Mouse Monoclonal Antibody [clone: DBC2/4570]
Catalog # 77180-784
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DBC 2
- Antigen Symbol:RHOBTB2
- Clonality:Monoclonal
- Clone:DBC2/4570
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9BYZ6
- Antigen Synonyms:Deleted in breast cancer 2|p83|DBC2|Deleted in breast cancer 2 gene protein|Rho related BTB domain containing protein 2|Rho related BTB domain containing 2|Rho-related BTB domain-containing protein 2|RHOBTB 2|RHBT2_HUMAN
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:83 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Human recombinant RHOBTB2 protein fragment (around Amino Acids 554-604). The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-784
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [DBC2/4570] antibody to RHOBTB2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: RHOBTB2
Clonality: Monoclonal
Clone: DBC2/4570
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human