Anti-PI 3 Kinase p110 delta Mouse Monoclonal Antibody [clone: PIK3CD/4639]
77180-716
- Antibody Type:Primary
- Antigen Name:5-bisphosphate 3-kinase 110 kDa catalytic subunit delta
- Antigen Symbol:PI 3 Kinase p110 delta
- Clonality:Monoclonal
- Clone:PIK3CD/4639
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00329
- Antigen Synonyms:Phosphatidylinositol 3 kinase catalytic delta polypeptide|APDS|IMD14|5-bisphosphate 3-kinase catalytic subunit delta isoform|p85-ALPHA|p110delta|p110d|p110dp85a|GRB1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:119 kDa/33 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human PIK3CD protein (around Amino Acids 520-720). The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-716
- Packaging:Plastic vial
Mouse monoclonal [PIK3CD/4639] antibody to PI 3 Kinase p110 delta for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: PI 3 Kinase p110 delta
Clonality: Monoclonal
Clone: PIK3CD/4639
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human