Anti-PHOX2B Rabbit Recombinant Antibody [clone: PHOX2B/7161R]
77180-712
- Antibody Type:Primary
- Antigen Name:NBLST2
- Antigen Symbol:PHOX2B
- Clonality:Recombinant
- Clone:PHOX2B/7161R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q99453
- Antigen Synonyms:PHOX 2B|Neuroblastoma paired type homeobox protein|PHOX 2B homeodomain protein|Paired-like homeobox 2B|Paired mesoderm homeobox protein 2B|Paired mesoderm homeobox 2b|Paired like homeobox 2b|NBPhox|Neuroblastoma Phox
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:32 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human PHOX2B protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-712
- Packaging:Plastic vial
Recombinant rabbit monoclonal [PHOX2B/7161R] antibody to PHOX2B for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: PHOX2B
Clonality: Recombinant
Clone: PHOX2B/7161R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human