Anti-Pet1 Mouse Monoclonal Antibody [clone: FEV/7311]
77180-700
- Antibody Type:Primary
- Antigen Name:ETS-domain transcription factor
- Antigen Symbol:Pet1
- Clonality:Monoclonal
- Clone:FEV/7311
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q99581
- Antigen Synonyms:FEV|FEV_HUMAN|mPet1|PC12 ETS domain-containing transcription factor 1|FEV ETS transcription factor|HSRNAFEV|Fifth Ewing sarcoma variant|FEV (ETS oncogene family)|Fifth Ewing variant protein
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:25 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human FEV protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-700
- Packaging:Plastic vial
Mouse monoclonal [FEV/7311] antibody to Pet1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Pet1
Clonality: Monoclonal
Clone: FEV/7311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human