Anti-PCNA Mouse Recombinant Antibody [clone: rPCNA/8859]
Catalog # 77180-648
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ATLD2
- Antigen Symbol:PCNA
- Clonality:Recombinant
- Clone:rPCNA/8859
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P12004
- Antigen Synonyms:DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:36 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human PCNA protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-648
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rPCNA/8859] antibody to PCNA for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: PCNA
Clonality: Recombinant
Clone: rPCNA/8859
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human