Anti-P Glycoprotein Rabbit Recombinant Antibody [clone: MDR1/8962R]
Catalog # 77180-544
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ABC20
- Antigen Symbol:P Glycoprotein
- Clonality:Recombinant
- Clone:MDR1/8962R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P08183
- Antigen Synonyms:Doxorubicin resistance|Colchicin sensitivity|ATP binding cassette, sub family B (MDR/TAP), member 1|CD243|MDR1|GP170|CLCS|ABCB1|ATP-binding cassette sub-family B member 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:170 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 500-700) of human ABCB1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-544
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [MDR1/8962R] antibody to P Glycoprotein for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: P Glycoprotein
Clonality: Recombinant
Clone: MDR1/8962R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human