Anti-Osteopontin Mouse Recombinant Antibody [clone: rOSP/8856]
77180-540
- Antibody Type:Primary
- Antigen Name:BNSP
- Antigen Symbol:Osteopontin
- Clonality:Recombinant
- Clone:rOSP/8856
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P10451
- Antigen Synonyms:Bone sialoprotein 1|BSP I|MGC110940|Nephropontin|ETA1|OPN|BSPI|Early T lymphocyte activation 1|ETA 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:34.9 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length rat Spp1 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-540
- Packaging:Plastic vial
Recombinant mouse monoclonal [rOSP/8856] antibody to Osteopontin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Osteopontin
Clonality: Recombinant
Clone: rOSP/8856
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human