Anti-NECAB1 Mouse Monoclonal Antibody [clone: NECAB1/7677]
Catalog # 77180-448
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:EF-hand calcium-binding protein 1
- Antigen Symbol:NECAB1
- Clonality:Monoclonal
- Clone:NECAB1/7677
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8N987
- Antigen Synonyms:EFCBP1|neuronal calcium binding protein|N-terminal EF-hand calcium-binding protein 1|synaptotagmin interacting protein 1|NECA1_HUMAN|STIP-1|Neuronal calcium-binding protein 1|NECAB1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:41 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human NECAB1 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-448
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [NECAB1/7677] antibody to NECAB1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: NECAB1
Clonality: Monoclonal
Clone: NECAB1/7677
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human