Anti-MR1 Mouse Monoclonal Antibody [clone: MR1/7577]
77180-338
- Antibody Type:Primary
- Antigen Name:Class I histocompatibility antigen like protein
- Antigen Symbol:MR1
- Clonality:Monoclonal
- Clone:MR1/7577
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q95460
- Antigen Synonyms:Major histocompatibility complex class I related isoform CRA d|Major histocompatibility complex class I related gene protein|Major histocompatibility complex class I like sequence|Major histocompatibility complex class I related isoform CRA a|Major histocompatibility complex class I related isoform CRA c|HMR1_HUMAN|Major histocompatibility complex class I related|Class I histocompatibility antigen-like protein|HLALS
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:39 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 50-250) of human MR1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-338
- Packaging:Plastic vial
Mouse monoclonal [MR1/7577] antibody to MR1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: MR1
Clonality: Monoclonal
Clone: MR1/7577
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human