Anti-Kallikrein 5 Mouse Monoclonal Antibody [clone: KLK5/4760]
Catalog # 77180-084
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:HK5
- Antigen Symbol:Kallikrein 5
- Clonality:Monoclonal
- Clone:KLK5/4760
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y337
- Antigen Synonyms:Kallikrein-5|Kallikrein like 2|Kallikrein like protein 2|Kallikrein5|Kallikrein related peptidase 5|Kallikrein 5|Kallikrein L2|Kallikrein-like protein 2|KLK 5
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:33 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human KLK5 protein (around Amino Acids 1-200). The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-084
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [KLK5/4760] antibody to Kallikrein 5 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Kallikrein 5
Clonality: Monoclonal
Clone: KLK5/4760
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human