Anti-Human IgD Rabbit Recombinant Antibody [clone: IGHD/6818R]
Catalog # 77179-954
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Human IgD
- Antigen Symbol:Human IgD
- Clonality:Recombinant
- Clone:IGHD/6818R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01880
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:44 - 80 kDa (heavy chain)/21 - 25 kDa (kappa light chain)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human IGHD protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-954
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [IGHD/6818R] antibody to Human IgD for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Human IgD
Clonality: Recombinant
Clone: IGHD/6818R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human