To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:EL52
- Antigen Symbol:HSP90 alpha
- Clonality:Monoclonal
- Clone:HSP90AA1/7247
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P07900
- Antigen Synonyms:Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:90 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 500-700) of human HSP90AA1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-912
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [HSP90AA1/7247] antibody to HSP90 alpha for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: HSP90 alpha
Clonality: Monoclonal
Clone: HSP90AA1/7247
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human