Anti-GIRK2 Mouse Monoclonal Antibody [clone: KCNJ6/7558]
Catalog # 77179-728
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BIR1
- Antigen Symbol:GIRK2
- Clonality:Monoclonal
- Clone:KCNJ6/7558
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P48051
- Antigen Synonyms:Inward rectifier potassium channel KIR3.2|inwardly rectifying subfamily J member 6|GIRK-2|G protein-activated inward rectifier potassium channel 2|GIRK2|G protein activated inward rectifier potassium channel 2|KATP-2|Inward rectifier K(+) channel Kir3.2|hiGIRK2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:48 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human KCNJ6 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-728
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [KCNJ6/7558] antibody to GIRK2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: GIRK2
Clonality: Monoclonal
Clone: KCNJ6/7558
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human