Anti-Gbx2 Mouse Monoclonal Antibody [clone: GBX2/7235]
77179-678
- Antibody Type:Primary
- Antigen Name:Gastrulation and brain-specific homeobox protein 2
- Antigen Symbol:Gbx2
- Clonality:Monoclonal
- Clone:GBX2/7235
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P52951
- Antigen Synonyms:Homeobox protein GBX-2|GBX2_HUMAN|Gastrulation brain homeobox 2|Homeobox protein STRA7|Mmoxa|Gastrulation brain homeo box 2|Homeobox protein GBX2|Homeobox protein GBX 2|Gbx2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:37 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 150-350) of human GBX2 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-678
- Packaging:Plastic vial
Mouse monoclonal [GBX2/7235] antibody to Gbx2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Gbx2
Clonality: Monoclonal
Clone: GBX2/7235
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human