Anti-FOXI1 Mouse Monoclonal Antibody [clone: PCRP-FOXI1-1C4]
Catalog # 77179-626
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:FKH10
- Antigen Symbol:FOXI1
- Clonality:Monoclonal
- Clone:PCRP-FOXI1-1C4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q12951
- Antigen Synonyms:FKHL10|Forkhead related activator 6|Forkhead box I1|Forkhead (Drosophila) like 10|Forkhead-related transcription factor 6|Forkhead-related protein FKHL10|Forkhead like 10|Forkhead related transcription factor 6|Forkhead box protein I1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:36 - 41 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 104-258) of human FOXL1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-626
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-FOXI1-1C4] antibody to FOXI1 for IP, Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated Applications: IP, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: IP: 1 to 2 µg per 100 to 500ug of total protein, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ ml, WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: FOXI1
Clonality: Monoclonal
Clone: PCRP-FOXI1-1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human