Anti-Factor VII Mouse Monoclonal Antibody [clone: F7/3515]
77179-578
- Antibody Type:Primary
- Antigen Name:Cf7
- Antigen Symbol:Factor VII
- Clonality:Monoclonal
- Clone:F7/3515
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P08709
- Antigen Synonyms:coagulation factor VII (serum prothrombin conversion accelerator)|F7 deficiency|Factor VII light chain|Eptacog alfa|Coagulation factor VII|FA7_HUMAN|FVII|Factor VII heavy chain|Factor VII deficiency
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:200 kDa/80 kDa/73 kDa/50 kDa/43 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 366-466) of human F7 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-578
- Packaging:Plastic vial
Mouse monoclonal [F7/3515] antibody to Factor VII for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Factor VII
Clonality: Monoclonal
Clone: F7/3515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human