Anti-Estrogen Receptor alpha Rabbit Recombinant Antibody [clone: ESR1/8206R]
77179-546
- Antibody Type:Primary
- Antigen Name:DKFZp686N23123
- Antigen Symbol:Estrogen Receptor alpha
- Clonality:Recombinant
- Clone:ESR1/8206R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P03372
- Antigen Synonyms:Estradiol receptor|ER|Era|ER alpha|ESRA|ER-alpha|ESR|ESR1|ESR1_HUMAN
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:~67 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 495-595) of human ER alpha protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Cat. No.:77179-546
- Packaging:Plastic vial
Recombinant rabbit monoclonal [ESR1/8206R] antibody to Estrogen Receptor alpha for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Recombinant
Clone: ESR1/8206R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human