Anti-Eph receptor B4 / HTK Mouse Monoclonal Antibody [clone: EPHB4/6391]
77179-514
- Antibody Type:Primary
- Antigen Name:Eph receptor B4 / HTK
- Antigen Symbol:Eph receptor B4 / HTK
- Clonality:Monoclonal
- Clone:EPHB4/6391
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human,Dog,Cat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P54760
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:120 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human EPHB4 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-514
- Packaging:Plastic vial
Mouse monoclonal [EPHB4/6391] antibody to Eph receptor B4 / HTK for IHC-P with samples derived from Human, Canine and Feline.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Eph receptor B4 / HTK
Clonality: Monoclonal
Clone: EPHB4/6391
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human;Dog;Cat