Order Entry
United States
ContactUsLinkComponent
 
Anti-ds DNA Rabbit Recombinant Antibody [clone: DSD/8204R]
  77179-476
Anti-ds DNA Rabbit Recombinant Antibody [clone: DSD/8204R]
  77179-476
 :  A316281-100

 

  • Antibody Type:
    Primary
  • Antigen Name:
    ds DNA
  • Antigen Symbol:
    ds DNA
  • Clonality:
    Recombinant
  • Clone:
    DSD/8204R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Liquid
  • Storage Buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Nuclei of Burkitt's cells.
  • Tested Applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Cat. No.:
    77179-476
  • Packaging:
    Plastic vial

 

 

Recombinant rabbit monoclonal [DSD/8204R] antibody to ds DNA for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: ds DNA
Clonality: Recombinant
Clone: DSD/8204R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human