Anti-Creatine Kinase MB Mouse Recombinant Antibody [clone: rCKBB/8842]
77179-322
- Antibody Type:Primary
- Antigen Name:B-CK
- Antigen Symbol:Creatine Kinase MB
- Clonality:Recombinant
- Clone:rCKBB/8842
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P12277
- Antigen Synonyms:creatine kinase muscle|Creatine Kinase MB|creatine kinase B|creatine kinase M|CKMM|brain creatine kinase|CKMB|creatine kinase brain|CKBB
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:42 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human full-length CKB protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-322
- Packaging:Plastic vial
Recombinant mouse monoclonal [rCKBB/8842] antibody to Creatine Kinase MB for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Creatine Kinase MB
Clonality: Recombinant
Clone: rCKBB/8842
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human