Anti-CPS1 Mouse Monoclonal Antibody [clone: CPS1/8420]
Catalog # 77179-308
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Carbamoyl phosphate synthase
- Antigen Symbol:CPS1
- Clonality:Monoclonal
- Clone:CPS1/8420
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P31327
- Antigen Synonyms:Carbamoyl-phosphate synthase [ammonia]|Carbamoylphosphate synthetase 1|Carbamoyl phosphate synthetase 1|Carbamoyl-phosphate synthetase I|Carbamoyl phosphate synthetase I|Carbamoyl phosphate synthase [ammonia]|Carbamoyl phosphate synthetase 1 mitochondrial|Carbamoylphosphate synthase|Carbamoyl phosphate synthase [ammonia] mitochondrial
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:~165 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human CPS1 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-308
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CPS1/8420] antibody to CPS1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CPS1
Clonality: Monoclonal
Clone: CPS1/8420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human