Anti-beta Actin Mouse Monoclonal Antibody [clone: ACTB/2370]
Catalog # 77178-690
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:A26C1A
- Antigen Symbol:beta Actin
- Clonality:Monoclonal
- Clone:ACTB/2370
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P60709
- Antigen Synonyms:Actin, cytoplasmic 1, N-terminally processed|b actin|ACTB|A26C1B|Actx|Actin beta|ACTB_HUMAN|Actin cytoplasmic 1|beta Actin
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:43 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 150-350) of human ACTB protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Cat. No.:77178-690
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ACTB/2370] antibody to beta Actin with samples derived from Human.
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: beta Actin
Clonality: Monoclonal
Clone: ACTB/2370
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human