Anti-Bcl6 Mouse Monoclonal Antibody [clone: PCRP-GCM2-1B3]
Catalog # 77178-660
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:B cell CLL/lymphoma 6
- Antigen Symbol:Bcl6
- Clonality:Monoclonal
- Clone:PCRP-GCM2-1B3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P41182
- Antigen Synonyms:B cell lymphoma 6 protein|Bcl 6|BCL 5|Bcl6|BCL5|BCL-5|B-cell lymphoma 6 protein|BCL-6|B-cell lymphoma 5 protein
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:56.61 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human GCM2 protein.
- Purification:Protein A/G chromatography
- Cat. No.:77178-660
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-GCM2-1B3] antibody to Bcl6 for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ ml
Type: Primary
Antigen: Bcl6
Clonality: Monoclonal
Clone: PCRP-GCM2-1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human