Anti-ARID1A Mouse Monoclonal Antibody [clone: ARID1A/7735]
Catalog # 77178-642
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:actin-dependent regulator of chromatin subfamily F member 1
- Antigen Symbol:ARID1A
- Clonality:Monoclonal
- Clone:ARID1A/7735
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O14497
- Antigen Synonyms:B120|AT rich interactive domain 1A (SWI like)|AT-rich interactive domain-containing protein 1A|ARID domain-containing protein 1A|AT rich interactive domain containing protein 1A|ARID domain containing protein 1A|AT rich interactive domain 1A|ARI1A_HUMAN|ARID1A
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:165 - 320 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1900-2000) of human ARID1A protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-642
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ARID1A/7735] antibody to ARID1A for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: ARID1A
Clonality: Monoclonal
Clone: ARID1A/7735
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human