Anti-MKP-1 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to MKP-1 for WB with samples derived from Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: MKP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77210-588
- Antigen Symbol:
- MKP-1
- Antigen Name:
- CL 100
- Antigen Synonyms:
- CL100|DUSP 1|Mitogen-activated protein kinase phosphatase 1|DUS1_HUMAN|MAP kinase phosphatase 1|Dual specificity protein phosphatase hVH1|Dual specificity protein phosphatase 1|Dual Specificity Phosphatase 1|DUSP1
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P28563
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant protein of mouse MKP1.
- Form:
- Liquid
- Molecular Weight:
- 40 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together