Anti-N6-methyladenosine / m6A Mouse Monoclonal Antibody [clone: AMC0257]
Supplier: Avantor
Certificates
About this item
Mouse monoclonal [AMC0257] antibody to N6-Methyladenosine / m6A for Dot Blot with samples derived from Species Independent.
- Validated Applications: Dot Blot
- Recommended Dilutions: DB: 1:500 to 1:2,000
Type: Primary
Antigen: N6-methyladenosine / m6A
Clonality: Monoclonal
Clone: AMC0257
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Species Independent
Related Information
Frequently Bought Together