Anti-MYO1B Rabbit Monoclonal Antibody [clone: ARC2990]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2990] antibody to MYO1B for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: MYO1B
Clonality: Monoclonal
Clone: ARC2990
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77216-826
- Antigen Symbol:
- MYO1B
- Antigen Name:
- MMI alpha
- Antigen Synonyms:
- Myosin I alpha|myosin IB|MYO1B variant protein|MMI-alpha|MYO1B_HUMAN|MYO1B|MMIa|MYH 1c|MYH-1c
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC2990
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# O43795
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant protein of Human MYO1B.
- Form:
- Liquid
- Molecular Weight:
- 132 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together