Anti-PAR4 Rabbit Monoclonal Antibody [clone: ARC1980]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1980] antibody to PAR4 for WB, IHC and ICC/IF with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: PAR4
Clonality: Monoclonal
Clone: ARC1980
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77203-580
- Antigen Symbol:
- PAR4
- Antigen Name:
- PAR4
- Antigen Synonyms:
- Pawr|PAR 4|Prostate apoptosis response protein 4|PRKC apoptosis WT1 regulator protein|2310001G03Rik|PAR4|PAR-4|PRKC Apoptosis WT1 Regulator|PAWR_HUMAN|Prostate apoptosis response 4 protein
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1980
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q96IZ0
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 241-340 of human PAR4 (Q96IZ0).
- Sequence:
- YSRTDRSGFPRYNRDANVSGTLVSSSTLEKKIEDLEKEVVRERQENLRLVRLMQDKEEMIGKLKEEIDLLNRDLDDIEDENEQLKQENKTLLKVVGQLTR
- Form:
- Liquid
- Molecular Weight:
- 41 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- IHC, ICC/IF
Specifications
Frequently Bought Together