Anti-PIGM Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to PIGM for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: PIGM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77202-040
- Antigen Symbol:
- PIGM
- Antigen Name:
- dol P Man dependent GPI mannosyltransferase
- Antigen Synonyms:
- GPI MT I|PIGM|Phosphatidylinositol glycan biosynthesis class M protein|PIG M mannosyltransferase|GPI mannosyltransferase I|DPM GlcN (acyl)PI mannosyltransferase|GPI mannosyltransferase 1|Phosphatidylinositol glycan anchor biosynthesis, class M
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9H3S5
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 35-96 of human PIGM (NP_660150.1).
- Sequence:
- YGVFQDRTLHVRYTDIDYQVFTDAARFVTEGRSPYLRATYRYTPLLGWLLTPNIYLSELFGK
- Form:
- Liquid
- Molecular Weight:
- 49 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- IHC, ICC/IF
Specifications
Frequently Bought Together