Anti-Uroplakin Ia Rabbit Recombinant Antibody [clone: UPK1A/8704R]
Catalog # 77178-456
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Uroplakin Ia
- Antigen Symbol:Uroplakin Ia
- Clonality:Recombinant
- Clone:UPK1A/8704R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00322
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:29 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 100-200) of human Uroplakin 1A (UPK1A) protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-456
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [UPK1A/8704R] antibody to Uroplakin Ia for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Uroplakin Ia
Clonality: Recombinant
Clone: UPK1A/8704R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human