Anti-Syndecan-1 Mouse Monoclonal Antibody [clone: SDC1/7177]
77178-330
- Antibody Type:Primary
- Antigen Name:CD138
- Antigen Symbol:Syndecan-1
- Clonality:Monoclonal
- Clone:SDC1/7177
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P18827
- Antigen Synonyms:heparan sulfate proteoglycan fibroblast growth factor receptor|SDC|Syndecan 1|Sdc1|Syndecan|SDC1_HUMAN|CD138 antigen|SYND1|CD_antigen
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:85 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-250) of human CD138 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-330
- Packaging:Plastic vial
Mouse monoclonal [SDC1/7177] antibody to Syndecan-1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Syndecan-1
Clonality: Monoclonal
Clone: SDC1/7177
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human