Anti-Stathmin 1 Mouse Monoclonal Antibody [clone: STMN1/8438]
Catalog # 77178-268
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:C1orf215
- Antigen Symbol:Stathmin 1
- Clonality:Monoclonal
- Clone:STMN1/8438
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P16949
- Antigen Synonyms:Op18|Oncoprotein 18|Metablastin|OP 18|LAP18|LAP 18|Lag|Leukemia-associated phosphoprotein p18|Leukemia associated phosphoprotein p18
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:19 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment of human STMN1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-268
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [STMN1/8438] antibody to Stathmin 1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Stathmin 1
Clonality: Monoclonal
Clone: STMN1/8438
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human