Anti-SHP1 Mouse Monoclonal Antibody [clone: PTPN6/7544]
Catalog # 77178-190
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:70Z-SHP
- Antigen Symbol:SHP1
- Clonality:Monoclonal
- Clone:PTPN6/7544
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P29350
- Antigen Synonyms:HPTP1C|HCPH|Hematopoietic cell protein-tyrosine phosphatase|Hematopoietic cell phosphatase|Protein tyrosine phosphatase non receptor type 6|Hematopoietic cell protein tyrosine phosphatase|HCP|Protein tyrosine phosphatase 1C|EC 3.1.3.48
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:65 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 395-595) of human PTPN6 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-190
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PTPN6/7544] antibody to SHP1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: SHP1
Clonality: Monoclonal
Clone: PTPN6/7544
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human