Anti-Osteopontin Mouse Recombinant Antibody [clone: rOSP/8856]
Catalog # 77177-772
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# P10451
- Antigen Synonyms:Bone sialoprotein 1|BSP I|MGC110940|Nephropontin|ETA1|OPN|BSPI|Early T lymphocyte activation 1|ETA 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:34.9 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length rat Spp1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77177-772
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rOSP/8856] antibody to Osteopontin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Osteopontin
Clonality: Recombinant
Clone: rOSP/8856
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human