Anti-LI Cadherin Mouse Recombinant Antibody [clone: rCDH17/8512]
Catalog # 77177-408
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# Q12864
- Antigen Synonyms:CDH16|CAD17_HUMAN|Cadherin-17|cadherin, liver-intestine|cadherin 17, LI cadherin (liver-intestine)|Cadherin 16, formerly|CDH16, formerly|Cadherin 16|Cadherin 17
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:120 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 242-418) of human Cadherin 17 protein (CDH17). The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:77177-408
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rCDH17/8512] antibody to LI Cadherin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: LI Cadherin
Clonality: Recombinant
Clone: rCDH17/8512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human