To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# P41229
- Antigen Synonyms:Lysine (K) specific demethylase 5C|Histone demethylase JARID1C|Jumonji/ARID domain-containing protein 1C|Jumonji, AT rich interactive domain 1C (RBP2 like)|KDM5C_HUMAN|JmjC domain containing protein SMCX|KDM5C|KDM5C / Jarid1C / SMCX|Jumonji AT rich interactive domain 1C
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:176 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human KDM5C protein
- Purification:Protein A/G chromatography
- Cat. No.:77177-334
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-KDM5C-1A11] antibody to KDM5C/Jarid1C/SMCX for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: KDM5C/Jarid1C/SMCX
Clonality: Monoclonal
Clone: PCRP-KDM5C-1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human