Anti-Integrin beta 1 Rabbit Recombinant Antibody [clone: ITGB1/8744R]
Catalog # 77177-298
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:beta1 integrin
- Antigen Symbol:Integrin beta 1
- Clonality:Recombinant
- Clone:ITGB1/8744R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05556
- Antigen Synonyms:FNRB|Glycoprotein IIa|Integrin beta 1|GP IIa|CD29|Integrin subunit beta 1|Integrin beta-1|GPIIA|Fibronectin receptor subunit beta
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:110 kDa (non-reduced)/130 kDa (reduced)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ITGB1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77177-298
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [ITGB1/8744R] antibody to Integrin beta 1 for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Integrin beta 1
Clonality: Recombinant
Clone: ITGB1/8744R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human