Anti-IGF2BP2/IMP-2 Mouse Monoclonal Antibody [clone: PCRP-IGF2BP2-1F9]
Catalog # 77177-194
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:IGF2BP2/IMP-2
- Clonality:Monoclonal
- Clone:PCRP-IGF2BP2-1F9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y6M1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:105 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human IGF2BP2 protein
- Purification:Protein A/G chromatography
- Cat. No.:77177-194
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-IGF2BP2-1F9] antibody to IGF2BP2/IMP-2 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: IGF2BP2/IMP-2
Clonality: Monoclonal
Clone: PCRP-IGF2BP2-1F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human