Anti-HIF-1 alpha Mouse Monoclonal Antibody [clone: HIF1A/3248]
77177-078
- Antibody Type:Primary
- Antigen Name:ARNT interacting protein
- Antigen Symbol:HIF-1 alpha
- Clonality:Monoclonal
- Clone:HIF1A/3248
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q16665
- Antigen Synonyms:HIF 1alpha|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|HIF-1-alpha|HIF-1 alpha|Basic helix loop helix PAS protein MOP1|bHLHe78|HIF 1A|Class E basic helix-loop-helix protein 78
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:92 - 110 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (aa 239-530) of human HIF-1 alpha protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77177-078
- Packaging:Plastic vial
Mouse monoclonal [HIF1A/3248] antibody to HIF-1 alpha for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: HIF-1 alpha
Clonality: Monoclonal
Clone: HIF1A/3248
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human