Anti-HDAC7 Mouse Monoclonal Antibody [clone: PCRP-HDAC7-1B6]
Catalog # 77177-054
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DKFZP586J0917
- Antigen Symbol:HDAC7
- Clonality:Monoclonal
- Clone:PCRP-HDAC7-1B6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8WUI4
- Antigen Synonyms:FLJ99588|HDAC7|HDAC7_HUMAN|HD7a|HDAC 7|HDAC 7A|HD7|HDAC7A|HD 7a
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:105 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human HDAC7 protein
- Purification:Protein A/G chromatography
- Cat. No.:77177-054
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-HDAC7-1B6] antibody to HDAC7 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: HDAC7
Clonality: Monoclonal
Clone: PCRP-HDAC7-1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human