To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# Q92896
- Antigen Synonyms:Cys -rich FGF Receptor|Cysteine rich fibroblast growth factor receptor|E selectin ligand 1|AW537898|CFR 1|DKFZp686L08213|CFR-1|Cysteine-rich fibroblast growth factor receptor|CFR1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:134 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human GLG1 protein
- Tested Applications:IHC
- Purification:Protein A/G chromatography
- Cat. No.:77176-996
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [GLG1/7174R] antibody to Golgi Complex for IHC with samples derived from Human.
- Validated Applications: IHC
- Recommended Dilutions: IHC: (Frozen and Formalin to fixed) 0.5 to 1 µg/ml
Type: Primary
Antigen: Golgi Complex
Clonality: Recombinant
Clone: GLG1/7174R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human