Anti-Glypican 3 Rabbit Recombinant Antibody [clone: GPC3/8363R]
Catalog # 77176-994
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# P51654
- Antigen Synonyms:Gpc3|Heparan sulphate proteoglycan|Glypican 3|Glypican-3 [Precursor]|Intestinal protein OCI 5|GTR2-2|GTR2 2|GPC3_HUMAN|Glypican proteoglycan 3
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:67 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 400-550) of human GPC3 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:77176-994
- Packaging:Plastic vial